Anti-DDX18 Rabbit polyclonal antibody unconjugated

Leverandør: Abcam
ABCAAB70526-100EA 11450 NOK
ABCAAB70526-100
Anti-DDX18 Rabbit polyclonal antibody unconjugated
Antistoffer
Type: Primary
Antigen: DDX18 (DEAD (Asp-Glu-Ala-Asp) box polypeptide 18)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR