Anti-EIF2S1 Rabbit polyclonal antibody unconjugated
Leverandør: Abcam
ABCAAB70542-100EA 11725 NOK
ABCAAB70542-100
Anti-EIF2S1 Rabbit polyclonal antibody unconjugated
Antistoffer
Type: Primary
Antigen: EIF2S1 (eukaryotic translation initiation factor 2, subunit 1 alpha, 35kDa)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse