Anti-BUB1B Rabbit Polyclonal Antibody
Leverandør: Abcam
ABCAAB70544-100EA 11725 NOK
ABCAAB70544-100
Anti-BUB1B Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: BUB1B (BUB1 mitotic checkpoint serine/threonine kinase B)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human