Anti-SKIV2L2 Rabbit Polyclonal Antibody
Leverandør: Abcam
ABCAAB70552-100EA 11400 NOK
ABCAAB70552-100
Anti-SKIV2L2 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: SKIV2L2 (superkiller viralicidic activity 2-like 2 (S. cerevisiae))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human