Anti-FANCE Mouse monoclonal antibody unconjugated [clone: 2346C5a]

ABCAAB70600-100EA 7550 NOK
ABCAAB70600-100
Anti-FANCE Mouse monoclonal antibody unconjugated [clone: 2346C5a]
Type: Primary
Antigen: FANCE (Fanconi anemia, complementation group E)
Clonality: monoclonal
Clone: 2346C5a
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR