Anti-TPR Rabbit Polyclonal Antibody
Leverandør: Abcam
ABCAAB70610-100EA 11475 NOK
ABCAAB70610-100
Anti-TPR Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: TPR (translocated promoter region, nuclear basket protein)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human