Anti-PPP4R1 Rabbit polyclonal antibody unconjugated

ABCAAB70624-100EA 7175 NOK
ABCAAB70624-100
Anti-PPP4R1 Rabbit polyclonal antibody unconjugated
Type: Primary
Antigen: PPP4R1 (protein phosphatase 4, regulatory subunit 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR