Anti-PAF1 Rabbit Polyclonal Antibody
Leverandør: Abcam
ABCAAB70638-100EA 11475 NOK
ABCAAB70638-100
Anti-PAF1 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: PAF1 (Paf1, RNA polymerase II associated factor, homolog (S. cerevisiae))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human