Anti-GAPDH Rabbit polyclonal antibody unconjugated

Leverandør: Abcam
ABCAAB70700-100EA 11475 NOK
ABCAAB70700-100
Anti-GAPDH Rabbit polyclonal antibody unconjugated
Antistoffer
Type: Primary
Antigen: GAPDH (glyceraldehyde-3-phosphate dehydrogenase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR