Anti-DLGAP5 Rabbit polyclonal antibody unconjugated

Leverandør: Abcam
ABCAAB70742-100EA 8450 NOK
ABCAAB70742-100
Anti-DLGAP5 Rabbit polyclonal antibody unconjugated
Antistoffer
Type: Primary
Antigen: DLGAP5 (discs, large (Drosophila) homolog-associated protein 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR