Anti-DHX29 Rabbit polyclonal antibody unconjugated

Leverandør: Abcam
ABCAAB70745-100EA 10925 NOK
ABCAAB70745-100
Anti-DHX29 Rabbit polyclonal antibody unconjugated
Antistoffer
Type: Primary
Antigen: DHX29 (DEAH (Asp-Glu-Ala-His) box polypeptide 29)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR