Anti-RCC1 Goat Polyclonal Antibody

Leverandør: Abcam
ABCAAB70749-100EA 11475 NOK
ABCAAB70749-100
Anti-RCC1 Goat Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: RCC1 (regulator of chromosome condensation 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR