Anti-RCC2 Rabbit Polyclonal Antibody
Leverandør: Abcam
ABCAAB70787-100EA 11450 NOK
ABCAAB70787-100
Anti-RCC2 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: RCC2 (regulator of chromosome condensation 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse