Anti-KIF20A Rabbit Polyclonal Antibody
Leverandør: Abcam
ABCAAB70791-100EA 11725 NOK
ABCAAB70791-100
Anti-KIF20A Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: KIF20A (kinesin family member 20A)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human