Anti-FCN1 Rabbit polyclonal antibody unconjugated
Leverandør: Abcam
ABCAAB70814-100EA 11700 NOK
ABCAAB70814-100
Anti-FCN1 Rabbit polyclonal antibody unconjugated
Antistoffer
Type: Primary
Antigen: FCN1 (ficolin (collagen/fibrinogen domain containing) 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat