Anti-TRAF1 Rabbit Polyclonal Antibody
Catalog # NOVUNB100-56170
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:TNF receptor-associated factor 1
- Antigen symbol:TRAF1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Western blot:Yes
- Gene ID:7185
- Antigen synonyms:MGC:10353|EBI6
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 μL
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: TRAF1 (TNF receptor-associated factor 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: