Anti-TRPM8 Rabbit Polyclonal Antibody (FITC (Fluorescein))
Catalog # NOVUNBP1-97311F
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:transient receptor potential cation channel, subfamily M, member 8
- Antigen symbol:TRPM8
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Rabbit
- ImmunoFluorescence:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Gene ID:79054
- Antigen synonyms:TRPP8|LTRPC6
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 μL
- Pk:0,1 mL
Specifications
About this item
Type: Primary
Antigen: TRPM8 (transient receptor potential cation channel, subfamily M, member 8)
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat