Anti-SIRT6 Rabbit Polyclonal Antibody
Leverandør: Thermo Fisher Scientific
PIEAPA116958EA 0 NOK
PIEAPA116958
Anti-SIRT6 Rabbit Polyclonal Antibody
Antistoffer
PA1-16958 is specific for the ~37 kDa SIRT6 protein. A non-specific higher molecular weight band can be seen at ~61 kDa of equal or greater intensity than the mSIRT6 band. Sirtuins regulate epigenetic gene silencing. SIRT6 (Mono-ADP-ribosyltransferase sirtuin-6), a mouse Sir2 homolog, is a chromatin-associated nuclear ADP-ribosyltransferase protein. It appears to be involved in DNA repair and may also play a role in human ageing.
Type: Primary
Antigen: SIRT6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat