Anti-IgG F(ab')2 Goat Polyclonal Antibody (Biotin)
Leverandør: Rockland Immunochemicals
ROCK610-1604EA 4025 NOK
ROCK610-1604
Anti-IgG F(ab')2 Goat Polyclonal Antibody (Biotin)
Antistoffer
This product has been assayed against 1.0 µg of Mouse IgG in a standard capture ELISA using Peroxidase Conjugated Streptavidin and ABTS (2,2’-azino-bis-(3-ethylbenthiazoline-6-sulfonic acid)) as a substrate for 30 minutes at room temperature.
Recommended Dilutions: ELISA: 1:20000 - 1:100000; Western Blot: 1:2000 - 1:10000; Immunochemistry: 1:1000 - 1:5000;
Restore with deionized water (or equivalent); 1.0 ml
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: F(ab')2
Host: Goat
Isotype: IgG
Reactivity: Mouse