Anti-ITGA4 Mouse Monoclonal Antibody [clone: 13B426]
Leverandør: United States Biological
USBI030348EA 7125 NOK
USBI030348
Anti-ITGA4 Mouse Monoclonal Antibody [clone: 13B426]
Antistoffer
Type: Primary
Antigen: ITGA4 (integrin, alpha 4 (antigen CD49D, alpha 4 subunit of VLA-4 receptor))
Clonality: Monoclonal
Clone: 13B426
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human