Anti-KDM3A Mouse Monoclonal Antibody [clone: 13B437]
Leverandør: United States Biological
USBI030359EA 6375 NOK
USBI030359
Anti-KDM3A Mouse Monoclonal Antibody [clone: 13B437]
Antistoffer
Type: Primary
Antigen: KDM3A (lysine (K)-specific demethylase 3A)
Clonality: Monoclonal
Clone: 13B437
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human