Anti-KLF15 Mouse Monoclonal Antibody [clone: 13B449]

Leverandør: United States Biological
USBI030371EA 6850 NOK
USBI030371
Anti-KLF15 Mouse Monoclonal Antibody [clone: 13B449]
Antistoffer
Type: Primary
Antigen: KLF15 (Kruppel-like factor 15)
Clonality: Monoclonal
Clone: 13B449
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR