Anti-KLHL25 Mouse Monoclonal Antibody [clone: 13B459]
Leverandør: United States Biological
USBI030381EA 6375 NOK
USBI030381
Anti-KLHL25 Mouse Monoclonal Antibody [clone: 13B459]
Antistoffer
Type: Primary
Antigen: KLHL25 (kelch-like family member 25)
Clonality: Monoclonal
Clone: 13B459
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human