Anti-KSHV ORF8 Rabbit Polyclonal Antibody [clone: 13B469]
Leverandør: United States Biological
USBI030391EA 6125 NOK
USBI030391
Anti-KSHV ORF8 Rabbit Polyclonal Antibody [clone: 13B469]
Antistoffer
Type: Primary
Antigen: KSHV ORF8
Clonality: Polyclonal
Clone: 13B469
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human