Anti-BPIFB1 Mouse Monoclonal Antibody [clone: 13B481]
Leverandør: United States Biological
USBI030403EA 7125 NOK
USBI030403
Anti-BPIFB1 Mouse Monoclonal Antibody [clone: 13B481]
Antistoffer
Type: Primary
Antigen: BPIFB1 (BPI fold containing family B, member 1)
Clonality: Monoclonal
Clone: 13B481
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human