Anti-BPIFB1 Mouse Monoclonal Antibody [clone: 13B482]
Leverandør: United States Biological
USBI030404EA 7125 NOK
USBI030404
Anti-BPIFB1 Mouse Monoclonal Antibody [clone: 13B482]
Antistoffer
Type: Primary
Antigen: BPIFB1 (BPI fold containing family B, member 1)
Clonality: Monoclonal
Clone: 13B482
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human