Anti-LRP5 Mouse Monoclonal Antibody [clone: 13B485]
Leverandør: United States Biological
USBI030407EA 7125 NOK
USBI030407
Anti-LRP5 Mouse Monoclonal Antibody [clone: 13B485]
Antistoffer
Type: Primary
Antigen: LRP5 (low density lipoprotein receptor-related protein 5)
Clonality: Monoclonal
Clone: 13B485
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human