Anti-CA2 Rabbit Polyclonal Antibody

Leverandør: US Biological
USBI033109EA 7100 NOK
USBI033109
Anti-CA2 Rabbit Polyclonal Antibody
Antistoffer
Recommended dilution: ELISA: 1:1000; Western Blot: 50-100

Type: Primary
Antigen: CA2 (carbonic anhydrase II)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR