Anti-CABP5 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Leverandør: US Biological
USBI033125-APCEA 10300 NOK
USBI033125-APC
Anti-CABP5 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Antistoffer
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!
Type: Primary
Antigen: CABP5 (calcium binding protein 5)
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human