Anti-CACNA2D3 Rabbit Polyclonal Antibody

Leverandør: US Biological
USBI033128EA 7100 NOK
USBI033128
Anti-CACNA2D3 Rabbit Polyclonal Antibody
Antistoffer
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500; Flow Cytometry: 1:10-50

Type: Primary
Antigen: CACNA2D3 (calcium channel, voltage-dependent, alpha 2/delta subunit 3)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR