Anti-ITPA Rabbit Polyclonal Antibody (Biotin)

Leverandør: US Biological
USBI037161-BIOTINEA 10300 NOK
USBI037161-BIOTIN
Anti-ITPA Rabbit Polyclonal Antibody (Biotin)
Antistoffer
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500

Type: Primary
Antigen: ITPA (inosine triphosphatase (nucleoside triphosphate pyrophosphatase))
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR