Anti-MMP26 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Leverandør: US Biological
USBI038509-APCEA 10300 NOK
USBI038509-APC
Anti-MMP26 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Antistoffer
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!
Type: Primary
Antigen: MMP26 (matrix metallopeptidase 26)
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human