Anti-NDOR1 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Leverandør: US Biological
USBI038845-FITCEA 10300 NOK
USBI038845-FITC
Anti-NDOR1 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antistoffer
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!
Type: Primary
Antigen: NDOR1 (NADPH dependent diflavin oxidoreductase 1)
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse