Anti-SH3RF2 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Leverandør: US Biological
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500
Type: Primary
Antigen: SH3RF2 (SH3 domain containing ring finger 2)
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human
BESTILL
Utvid alle / Skjul alle
Str Fp Art.nr Antall
200 µl 200 µl USBI041696-HRP
Lag barcode etiketter Barcode etikett format: 
Lag PDF katalog
SPESIFIKASJONER
Antigen symbol SH3RF2
Antigen name SH3 domain containing ring finger 2
Reaktivitet Human
Konjugasjon HRP (Horseradish Peroxidase)
Klonalitet Polyclonal
antistofftype Primary
Vert Rabbit
Isotype IgG
Western blot Yes
ELISA Yes
aminosyre nr 456 to 483
Epitope internal
Rensing purified by Protein A chromatography
Form liquid
Lagringsbuffer PBS, pH 7,2
Lagringstemp Cool
Frakttemp. Ice
Legg inn detaljer
Velg varen hvor du søker etter sertifikat
Legg inn batchnummer. Legg inn "X" hvis du ikke har informasjonen for VWR Chemicals. Feltet er obligatorisk
Skriv inn serifikatdetaljer
Skriv inn lot nummer:
Velg fra de siste batch / lot tallene:
Eller
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beskrivelse Art.nr. Availability Enhet Din pris Price Per Antall