Anti-TXK Goat Polyclonal Antibody

Leverandør: United States Biological
USBI149881EA 9050 NOK
USBI149881
Anti-TXK Goat Polyclonal Antibody
Antistoffer
Recommended dilution: ELISA: 1:4000; Western Blot: 2-4?g/ml

Type: Primary
Antigen: TXK (TXK tyrosine kinase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR