Anti-MPO Rabbit Polyclonal Antibody

Leverandør: United States Biological
USBI168381EA 11000 NOK
USBI168381
Anti-MPO Rabbit Polyclonal Antibody
Antistoffer
Recommended dilution: ELISA: 1:5000-1:20000; Western Blot: 1:500-1:5000; Immunoprecipitation: 1:100

Type: Primary
Antigen: MPO (myeloperoxidase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR