Anti-DIABLO Rabbit polyclonal antibody

Leverandør: United States Biological
USBIS1014-48FEA 5725 NOK
USBIS1014-48F
Anti-DIABLO Rabbit polyclonal antibody
Antistoffer
Type: Primary
Antigen: DIABLO (diablo, IAP-binding mitochondrial protein)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR