Anti-DIABLO Rabbit polyclonal antibody
Leverandør: United States Biological
USBIS1014-48JEA 5300 NOK
USBIS1014-48J
Anti-DIABLO Rabbit polyclonal antibody
Antistoffer
Recommended dilution: Western Blot: 1:1000-1:2000; Immunochemistry: 1:1000-1:5000; Immunoprecipitation: 1:50-1:200
Type: Primary
Antigen: DIABLO (diablo, IAP-binding mitochondrial protein)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human