Anti-DIABLO Mouse monoclonal antibody [clone: 79-1-83]
Leverandør: United States Biological
USBIS1014-48VEA 8175 NOK
USBIS1014-48V
Anti-DIABLO Mouse monoclonal antibody [clone: 79-1-83]
Antistoffer
Recommended dilution: Immunochemistry: 10 ?g/ml
Type: Primary
Antigen: DIABLO (diablo, IAP-binding mitochondrial protein)
Clonality: Monoclonal
Clone: 79-1-83
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Monkey