Anti-DIABLO Rabbit polyclonal antibody

USBIS1014-48XEA 3675 NOK
USBIS1014-48X
Anti-DIABLO Rabbit polyclonal antibody
Recommended dilution: Immunochemistry: 5 μg/ml

Type: Primary
Antigen: DIABLO (diablo, IAP-binding mitochondrial protein)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR