Anti-SREBF1 Rabbit Polyclonal Antibody (FITC (Fluorescein))
Leverandør: United States Biological
USBIS6505-15D-FITCEA 10300 NOK
USBIS6505-15D-FITC
Anti-SREBF1 Rabbit Polyclonal Antibody (FITC (Fluorescein))
Antistoffer
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-1:500
Do not freeze!
Type: Primary
Antigen: SREBF1 (sterol regulatory element binding transcription factor 1)
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human