Anti-S Tag Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: 5K159]
Leverandør: United States Biological
USBIS6609-95FEA 8075 NOK
USBIS6609-95F
Anti-S Tag Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: 5K159]
Antistoffer
Type: Tag
Antigen: S Tag (S tag)
Clonality: Monoclonal
Clone: 5K159
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity:
Bestill nå