Norway
Order Entry
Anti-IgG Donkey Polyclonal Antibody (Biotin (NHS-LC))
  IMMRIR1254
Anti-IgG Donkey Polyclonal Antibody (Biotin (NHS-LC))
  IMMRIR1254
 
kr 2 850,00
   
  1 * 1 mg

 

  • Antibody type:
    Secondary
  • Antigen name:
    Immunoglobulin G
  • Antigen symbol:
    IgG
  • Clonality:
    Polyclonal
  • Conjugation:
    Biotin (NHS-LC)
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • Host:
    Donkey
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Rat
  • Western blot:
    Yes
  • Cross adsorption:
    Yes
  • Format:
    Purified IgG
  • Form:
    Clear, colorless liquid
  • Storage buffer:
    10 mM Sodium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 1% (w/v) BSA, Protease/IgG free
  • Storage temperature:
    2...8 °C
  • Concentration:
    1,50 mg/ml (E 1% at 280 nm = 13,0)
  • Shipping temperature:
    2...8 °C
  • Immunogen:
    Rat IgG
  • Tested applications:
    Fluorescence-Activated Cell Sorting
  • Purification:
    Affinity purified using solid phaseRat IgG
  • Size:
    2,0 mg
  • Pk:
    1 mg