Culture Media, Ready To Use, In Plates
New Product
A complete range of ready-to-use Petri plates for various microbiological applications.
available in 60 mm, 90 mm and 140 mm diameters
- most of the plates can be transported and stored at room temperature
- StandardsFillingPk
- Agar Burkholderia cepacia selective USP 90 mmAgar Burkholderia cepacia selective USP 90 mm10460——201 * 20 Itemskr 670,00: 10460: LIOF10460
- Bile Aesculin azide agarBile Aesculin azide agar163572ISO 7899-2—201 * 20 Itemskr 410,00: 163572: LIOF163572
- Brillant green agarBrillant green agarLIOF1002210022—22 ml201 * 20 Itemskr 350,00: 10022: LIOF10022
- Buffered petone with TSABuffered petone with TSALIOF1003710037EP—201 * 20 Itemskr 340,00: 10037: LIOF10037
- Chocolate agar 90 mmChocolate agar 90 mmLIOF1002310023——201 * 20 Itemskr 470,00: 10023: LIOF10023
- Chromatic Bacillus cereus 90 mmChromatic Bacillus cereus 90 mmLIOF11628X11628X——1001 * 100 Itemskr 3 050,00: 11628X: LIOF11628X
- Chromatic Bacillus cereus 90 mmChromatic Bacillus cereus 90 mmLIOF1162811628——201 * 20 Itemskr 730,00: 11628: LIOF11628
- Chromatic Clostridium difficile 90 mmChromatic Clostridium difficile 90 mmLIOF1163211632——201 * 20 Itemskr 1 400,00: 11632: LIOF11632
- Chromatic Coliform agar ISO 90 mmChromatic Coliform agar ISO 90 mmLIOF1163011630——201 * 20 Itemskr 1 150,00: 11630: LIOF11630
- Chromatic Colistin 90 mmChromatic Colistin 90 mmLIOF1164011640——201 * 20 Itemskr 1 150,00: 11640: LIOF11640
- Chromatic CRE 90 mmChromatic CRE 90 mmLIOF1161911619——201 * 20 Itemskr 820,00: 11619: LIOF11619
- Chromatic Cronobacter isolation agar vChromatic Cronobacter isolation agar v11636——201 * 20 Itemskr 1 275,00: 11636: LIOF11636
- Chromatic enterococcus 90 mmChromatic enterococcus 90 mmLIOF1162711627——201 * 20 Itemskr 1 200,00: 11627: LIOF11627
- Chromatic ESBL 90 mmChromatic ESBL 90 mmLIOF1162211622——201 * 20 Itemskr 1 100,00: 11622: LIOF11622
- Chromatic Esbl V. 90 mmChromatic Esbl V. 90 mmLIOF11626X11626X——1001 * 100 Itemskr 4 850,00: 11626X: LIOF11626X
- Chromatic ESBL V.C. 90 mmChromatic ESBL V.C. 90 mmLIOF11624X11624X——1001 * 100 Itemskr 4 850,00: 11624X: LIOF11624X
- Chromatic Esbl V.E. 90 mmChromatic Esbl V.E. 90 mmLIOF11625X11625X——1001 * 100 Itemskr 4 850,00: 11625X: LIOF11625X
- Chromatic GBS 20 90 mmChromatic GBS 20 90 mmLIOF1163911639——201 * 20 Itemskr 1 400,00: 11639: LIOF11639
- Chromatic MH 90 mmChromatic MH 90 mmLIOF1161811618——201 * 20 Itemskr 1 100,00: 11618: LIOF11618
- Chromatic MRSA 90 mmChromatic MRSA 90 mmLIOF1059910599——201 * 20 Itemskr 1 150,00: 10599: LIOF10599
- Chromatic OXA-48 90 mmChromatic OXA-48 90 mmLIOF1163111631——201 * 20 Itemskr 1 650,00: 11631: LIOF11631
- Chromatic Vibrio 90 mmChromatic Vibrio 90 mmLIOF1163311633——201 * 20 Itemskr 1 000,00: 11633: LIOF11633
- Chromatic VRE 90 mmChromatic VRE 90 mmLIOF1162111621——201 * 20 Itemskr 1 100,00: 11621: LIOF11621
- Chromatic™ SalmonellaChromatic™ SalmonellaLIOF1161411614——201 * 20 Itemskr 675,00: 11614: LIOF11614
- Coliform: 11183: LIOF11183
- Dichloran rose bengal agarDichloran rose bengal agarLIOF1009710097—22 ml11 * 1 Itemskr 620,00: 10097: LIOF10097
- EC MF agarEC MF agarLIOF163862163862——201 * 20 Itemskr 660,00: 163862: LIOF163862
- EMB Levine agarEMB Levine agarLIOF1004810048USP20 ml201 * 20 Itemskr 385,00: 10048: LIOF10048
- Endo agar: 10010: LIOF10010
- Enterobacter (Cronobacter) Sakazakii ISO 90 mmEnterobacter (Cronobacter) Sakazakii ISO 90 mmLIOF1162311623——201 * 20 Itemskr 865,00: 11623: LIOF11623
- Fecaux: 11210: LIOF11210
- Fecaux: 10057: LIOF10057
- Legionella agar (BCYE), 60 mmLegionella agar (BCYE), 60 mmLIOF163362163362——201 * 20 Itemskr 715,00: 163362: LIOF163362New ProductNew Product
- Legionella agar (GVPC), 60 mm: 163392: LIOF163392New ProductNew Product
- Legionella MWY selective agarLegionella MWY selective agarLIOF1012710127ISO 11731-1—201 * 20 Itemskr 810,00: 10127: LIOF10127
- Listeria aesculin agarListeria aesculin agarLIOF1123011230ISO 11290-1—201 * 20 Itemskr 410,00: 11230: LIOF11230
- Listeria agarListeria agarLIOF1062010620ISO 11290-1—201 * 20 Itemskr 1 025,00: 10620: LIOF10620
- MacConkey brothMacConkey brothLIOF2012620126——101 * 10 Itemskr 250,00: 20126: LIOF20126
- M-Endo agarM-Endo agarLIOF163432163432APHA—201 * 20 Itemskr 425,00: 163432: LIOF163432
- Mannitol salt agarMannitol salt agar10030USP/EP/JP—201 * 20 Itemskr 335,00: 10030: LIOF10030
- M-Endo agarM-Endo agar11053—20 ml201 * 20 Itemskr 515,00: 11053: LIOF11053
- M-Green yeast and mold agar pH modified: 163842L: LIOF163842L
- Nutrient agar ISO 6579 plates 90 mmNutrient agar ISO 6579 plates 90 mmLIOF1151311513——201 * 20 Itemskr 410,00: 11513: LIOF11513
- O.A. Listeria agar 90 mmO.A. Listeria agar 90 mmLIOF1022310223——201 * 20 Itemskr 2 125,00: 10223: LIOF10223
- PCA Plate count agarPCA Plate count agar10032ISO 483322 ml201 * 20 Itemskr 340,00: 10032: LIOF10032
- Pseudomonas CN agar plates 90 mmPseudomonas CN agar plates 90 mmLIOF1103411034——201 * 20 Itemskr 605,00: 11034: LIOF11034
- Sabouraud agarSabouraud agarLIOF10035/S10035/SEP22 ml201 * 20 Itemskr 540,00: 10035/S: LIOF10035/S
- Salmonella agarSalmonella agarLIOF1003610036—22 ml201 * 20 Itemskr 340,00: 10036: LIOF10036
- Tryptic soy agar and neutralising irradiated 90 mmTryptic soy agar and neutralising irradiated 90 mmLIOF10074S10074S——201 * 20 Itemskr 755,00: 10074S: LIOF10074S
- Tryptic Soy Agar Horse Blood 5%Tryptic Soy Agar Horse Blood 5%LIOF1103711037——11 * 1 Itemskr 495,00: 11037: LIOF11037
- Tryptic Soy Agar Horse Blood 5%Tryptic Soy Agar Horse Blood 5%LIOF1103811038——201 * 20 Itemskr 480,00: 11038: LIOF11038
- Tryptic soy agar irradiated 90 mmTryptic soy agar irradiated 90 mm10037S——201 * 20 Itemskr 610,00: 10037S: LIOF10037S
- Tryptone glucose extract agar + TTC: 163742L: LIOF163742L
- Vogel Johnson agarVogel Johnson agar11185USP—201 * 20 Itemskr 395,00: 11185: LIOF11185
- VRBLA + mug plate 90 mmVRBLA + mug plate 90 mmLIOF1001210012——201 * 20 Itemskr 530,00: 10012: LIOF10012
- XLD agarXLD agar10413EP, USP, JP—201 * 20 Itemskr 570,00: 10413: LIOF10413
- Yeast extract agar: 163582: LIOF163582New ProductNew Product