Norway
Order Entry
VWR®, Gel Cutters and Safety Razors
VWR®, Gel Cutters and Safety Razors
Supplier:  VWR Collection
Certificates
undefined
VWR®, Gel Cutters and Safety Razors
Supplier:  VWR Collection
Product Information
  • Catalog #
    Material
    Thickness
    L×W
    Pk
    Availability
    Price
  • Gel cutter and safety razor, regular
    Gel cutter and safety razor, regular
    700-0417
    PET
    0,27 mm
    25×19 mm
    100
    Log in for availability
    Log in for availability
    1 * 100 Items
    1 * 100 Items - kr 395,00
    kr 395,00
    1 * 100 Items
       kr 395,00
    Local Article Number: VWRI700-0417
    Specifications:
    Specifications
    Environmentally Preferable
  • Gel cutter and safety razor, large
    Gel cutter and safety razor, large
    700-0418
    PET
    0,27 mm
    50×25 mm
    100
    Log in for availability
    Log in for availability
    1 * 100 Items
    1 * 100 Items - kr 445,00
    kr 445,00
    1 * 100 Items
       kr 445,00
    Local Article Number: VWRI700-0418
    Specifications:
    Specifications
    Environmentally Preferable
Frequently Bought Together