Anti-ARFGAP3 Rabbit Polyclonal Antibody

Leverandør: OriGene
ORIGTA319362EA 9925 NOK
ORIGTA319362
Anti-ARFGAP3 Rabbit Polyclonal Antibody
Antistoffer
Recommended Dilutions: ELISA: 1:5000; Western Blot: 1:1000; Immunofluorescence: 1:100

Type: Primary
Antigen: ARFGAP3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR