Anti-ABCB1 Mouse Monoclonal Antibody [clone: 11D2]

Leverandør: OriGene
ORIGCF806689EA 8800 NOK
ORIGCF806689
Anti-ABCB1 Mouse Monoclonal Antibody [clone: 11D2]
Antistoffer
Recommended Dilutions: Immunochemistry: 1:150

Type: Primary
Antigen: ABCB1
Clonality: Monoclonal
Clone: 11D2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR