Anti-POSTN Mouse Monoclonal Antibody [clone: 2F7]

Leverandør: OriGene
ORIGCF803712EA 8800 NOK
ORIGCF803712
Anti-POSTN Mouse Monoclonal Antibody [clone: 2F7]
Antistoffer
Recommended Dilutions: Western Blot: 1:500; Immunochemistry: 1:150

Type: Primary
Antigen: POSTN
Clonality: Monoclonal
Clone: 2F7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR