Anti-RNA polymerase II subunit B1 Rabbit Polyclonal Antibody

Leverandør: Agrisera
AGRIAS111804EA 3675 NOK
AGRIAS111804
Anti-RNA polymerase II subunit B1 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: RNA polymerase II subunit B1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: A. thaliana
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR