Norway
Order Entry
Sample vials, with cap
Sample vials, with cap
undefined
Sample vials, with cap
  • Catalog No:
  • EPSCT346-0040
  • EPSCT336-0040
  • EPSCT236-0040
  • EPSCT136-0040
  • EPSCT246-0040
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Environmental certification:
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  •  
    Type
    Capacity
    Ø×H
    Colour
    Cap included
  • Unprocessed
    Unprocessed
    EPSCT136-0040
    T136-0040
    Unprocessed, open-top cap with 1,52 mm bonded septa
    40,00 ml
    28×95 mm
    Clear
    Yes
    1 * 72 Items
    1 * 72 Items - kr 4 250,00
    kr 4 250,00
    1 * 72 Items
       kr 4 250,00
     : T136-0040
     : EPSCT136-0040
      Low Manufacturing Impact
    Specifications
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • VOA processed
    VOA processed
    EPSCT236-0040
    T236-0040
    VOA processed, open-top cap with 1,52 mm bonded septa
    40,00 ml
    28×95 mm
    Clear
    Yes
    1 * 72 Items
    1 * 72 Items - kr 6 150,00
    kr 6 150,00
    1 * 72 Items
       kr 6 150,00
     : T236-0040
     : EPSCT236-0040
      Low Manufacturing Impact
    Specifications
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • VOA certified
    VOA certified
    EPSCT336-0040
    T336-0040
    VOA certified, open-top cap with 1,52 mm bonded septa
    40,00 ml
    28×95 mm
    Clear
    Yes
    1 * 72 Items
    1 * 72 Items - kr 7 350,00
    kr 7 350,00
    1 * 72 Items
       kr 7 350,00
     : T336-0040
     : EPSCT336-0040
      Low Manufacturing Impact
    Specifications
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • VOA processed
    VOA processed
    EPSCT246-0040
    T246-0040
    VOA processed, open-top cap with 1,52 mm bonded septa
    40,00 ml
    28×95 mm
    Amber
    Yes
    1 * 72 Items
    1 * 72 Items - kr 6 150,00
    kr 6 150,00
    1 * 72 Items
       kr 6 150,00
     : T246-0040
     : EPSCT246-0040
      Low Manufacturing Impact
    Specifications
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • VOA certified
    VOA certified
    EPSCT346-0040
    T346-0040
    VOA certified, open-top cap with 1,52 mm bonded septa
    40,00 ml
    28×95 mm
    Amber
    Yes
    1 * 72 Items
    1 * 72 Items - kr 7 350,00
    kr 7 350,00
    1 * 72 Items
       kr 7 350,00
     : T346-0040
     : EPSCT346-0040
      Low Manufacturing Impact
    Specifications
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact